Assay Details
| Gene Symbol: Vti1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293685.1 | NP_001280614.1 | 3 - 4 | 663 | 90 | |
| NM_001293686.1 | NP_001280615.1 | 3 - 4 | 663 | 90 | ||
| NM_016862.4 | NP_058558.1 | 3 - 4 | 663 | 90 | ||
| GenBank mRNA | AF035209.1 | - | 3 - 4 | 370 | 90 | |
| AF035823.1 | - | 3 - 4 | 368 | 90 | ||
| AK004751.1 | - | 3 - 4 | 359 | 90 | ||
| AK028646.1 | - | 3 - 4 | 369 | 90 | ||
| AK030538.1 | - | 3 - 4 | 388 | 90 | ||
| AK135463.1 | - | 3 - 4 | 426 | 90 | ||
| AK148281.1 | - | 3 - 4 | 394 | 90 | ||
| BC012211.1 | - | 3 - 4 | 90 | |||
| BC019386.1 | - | 3 - 4 | 5012235 | 90 | ||
| BC089321.1 | - | 3 - 4 | 370 | 6839437 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Vti1a
Genomic Map