Assay Details
| Gene Symbol: Idh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111320.1 | NP_001104790.1 | 1 - 2 | 173 | 85 | |
| GenBank mRNA | CF950262.1 | - | 1 - 2 | 112 | 85 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001111320.1 transcripts.
Set Membership: |
|

Idh1
Genomic Map