Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1800009907.mm.1 |
NM_175751.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800009907.mm.1 |
NM_175751.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800009907.mm.1 |
TC1800001345.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1800009905.mm.1 |
NM_175751.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800009905.mm.1 |
NM_175751.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800009905.mm.1 |
TC1800001345.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1800009906.mm.1 |
NM_175751.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800009906.mm.1 |
NM_175751.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800009906.mm.1 |
TC1800001345.mm.1 |
MTA TC |