Assay Details
| Gene Symbol: Slc14a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110273.1 | NP_001103743.1 | 7 - 8 | 1573 | 77 | |
| NM_207651.3 | NP_997534.2 | 18 - 19 | 2713 | 77 | ||
| GenBank mRNA | AF366052.2 | - | 18 - 19 | 2705 | 77 | |
| AF367359.1 | - | 7 - 8 | 1534 | 77 | ||
| AK032837.1 | - | 9 - 10 | 1317 | 77 | ||
| AY221737.1 | - | 4 - 5 | 410 | 77 | ||
| BC150680.1 | - | 18 - 19 | 2621 | 9087591 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc14a2
Genomic Map