Assay Details
Gene Symbol: Txnl4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042408.1 | NP_001035867.1 | 1 - 2 | 81 | 123 | |
GenBank mRNA | BC014770.1 | - | 1 - 2 | 81 | 123 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC014770.1, NM_001042408.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
