Assay Details
| Gene Symbol: Myl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113387.1 | NP_001106858.1 | 2 - 3 | 114 | 88 | |
| GenBank mRNA | AK009938.1 | - | 2 - 3 | 103 | 88 | |
| BC059087.1 | - | 2 - 3 | 81 | 30139109 | 88 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK009938.1, BC059087.1, NM_001113387.1 transcripts.
Set Membership: |
|

Myl1
Genomic Map