Assay Details
| Gene Symbol: Cyp2j12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100182.2 | NP_001093652.1 | 2 - 3 | 486 | 127 | |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cyp2j12| Gene Symbol: Cyp2j12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100182.2 | NP_001093652.1 | 2 - 3 | 486 | 127 | |
Set Membership: |
|