Assay Details
| Gene Symbol: Pip5k1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293707.1 | NP_001280636.1 | 11 - 12 | 1804 | 79 | |
| NM_008847.3 | NP_032873.2 | 11 - 12 | 1801 | 79 | ||
| GenBank mRNA | AK098097.1 | - | 11 - 12 | 1758 | 79 | |
| AK150898.1 | - | 11 - 12 | 1761 | 79 | ||
| AK151984.1 | - | 11 - 12 | 1761 | 79 | ||
| AK158062.1 | - | 11 - 12 | 1746 | 79 | ||
| BC031774.1 | - | 11 - 12 | 1729 | 4503697 | 79 | |
| D86177.1 | - | 11 - 12 | 1591 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pip5k1a
Genomic Map