Assay Details
| Gene Symbol: Pip5k1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293707.1 | NP_001280636.1 | 9 - 10 | 1662 | 77 | |
| NM_008847.3 | NP_032873.2 | 9 - 10 | 1659 | 77 | ||
| GenBank mRNA | AK098097.1 | - | 9 - 10 | 1616 | 77 | |
| AK150898.1 | - | 9 - 10 | 1619 | 77 | ||
| AK151984.1 | - | 9 - 10 | 1619 | 77 | ||
| AK158062.1 | - | 9 - 10 | 1604 | 77 | ||
| BC031774.1 | - | 9 - 10 | 1587 | 4503697 | 77 | |
| D86177.1 | - | 9 - 10 | 1449 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pip5k1a
Genomic Map