Assay Details
Gene Symbol: Abi2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198570.1 | NP_001185499.1 | 7 - 8 | 1027 | 97 | |
GenBank mRNA | BC079646.1 | - | 7 - 8 | 951 | 6850720 | 97 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
