Assay Details
| Gene Symbol: Ccbe1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178793.4 | NP_848908.1 | 6 - 7 | 749 | 58 | |
| GenBank mRNA | AK028377.1 | - | 6 - 7 | 747 | 58 | |
| AK035153.1 | - | 6 - 7 | 744 | 58 | ||
| AK039742.1 | - | 6 - 7 | 679 | 58 | ||
| AK087673.1 | - | 767 | 58 | |||
| AK220435.1 | - | 6 - 7 | 719 | 58 | ||
| BC103803.1 | - | 6 - 7 | 699 | 30088062 | 58 | |
| BC152322.1 | - | 6 - 7 | 718 | 40140631 | 58 | |
More Information
Set Membership: |
|

Ccbe1
Genomic Map