Assay Details
| Gene Symbol: Csnk1a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK078398.1 | - | 9 - 10 | 1425 | 73 | |
| AK146873.1 | - | 9 - 10 | 1446 | 73 | ||
| AK152793.1 | - | 9 - 10 | 1441 | 73 | ||
| AK167550.1 | - | 9 - 10 | 1445 | 73 | ||
| AK217167.1 | - | 1 - 2 | 165 | 73 | ||
| BC002171.1 | - | 3 - 4 | 324 | 73 | ||
More Information
Set Membership: |
|

Csnk1a1
Genomic Map