Assay Details
| Gene Symbol: Txnl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016792.4 | NP_058072.2 | 1 - 2 | 286 | 92 | |
| GenBank mRNA | AF052660.1 | - | 1 - 2 | 115 | 92 | |
| AK029807.1 | - | 1 - 2 | 287 | 92 | ||
| AK051698.1 | - | 1 - 2 | 261 | 92 | ||
| AK144211.1 | - | 1 - 2 | 243 | 92 | ||
| AK150326.1 | - | 1 - 2 | 249 | 92 | ||
| AK150887.1 | - | 1 - 2 | 255 | 92 | ||
| AK150983.1 | - | 1 - 2 | 255 | 92 | ||
| AK152101.1 | - | 1 - 2 | 255 | 92 | ||
| AK168712.1 | - | 1 - 2 | 250 | 92 | ||
| BC061123.1 | - | 1 - 2 | 313 | 6596299 | 92 | |
More Information
Set Membership: |
|

Txnl1
Genomic Map