Assay Details
| Gene Symbol: Nars | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142950.1 | NP_001136422.1 | 1 - 2 | 140 | 81 | |
| GenBank mRNA | AK083730.1 | - | 1 - 2 | 97 | 81 | |
| AK157244.1 | - | 1 - 2 | 99 | 81 | ||
| AK172657.1 | - | 1 - 2 | 99 | 81 | ||
| BC052849.1 | - | 1 - 2 | 98 | 30057822 | 81 | |
| BY117114.1 | - | 1 - 2 | 141 | 81 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK172657.1, BC052849.1, NM_001142950.1 transcripts.
Set Membership: |
|

Nars
Genomic Map