Assay Details
| Gene Symbol: Nars | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142950.1 | NP_001136422.1 | 5 - 6 | 471 | 103 | |
| NM_027350.3 | NP_081626.2 | 5 - 6 | 468 | 103 | ||
| GenBank mRNA | AK077699.1 | - | 5 - 6 | 449 | 103 | |
| AK157244.1 | - | 5 - 6 | 430 | 103 | ||
| AK172657.1 | - | 5 - 6 | 430 | 103 | ||
| BC005473.1 | - | 5 - 6 | 374 | 103 | ||
| BC052849.1 | - | 5 - 6 | 429 | 30057822 | 103 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nars
Genomic Map