Assay Details
| Gene Symbol: Nars | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142950.1 | NP_001136422.1 | 9 - 10 | 934 | 82 | |
| NM_027350.3 | NP_081626.2 | 9 - 10 | 931 | 82 | ||
| GenBank mRNA | AK077699.1 | - | 9 - 10 | 912 | 82 | |
| AK157244.1 | - | 9 - 10 | 894 | 82 | ||
| AK172657.1 | - | 9 - 10 | 893 | 82 | ||
| BC005473.1 | - | 9 - 10 | 837 | 82 | ||
| BC052849.1 | - | 9 - 10 | 892 | 30057822 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nars
Genomic Map