| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1800004791.mm.1 |
NM_028017.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800004791.mm.1 |
NM_028017.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800004791.mm.1 |
TC1800000648.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1800004792.mm.1 |
NM_028017.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800004792.mm.1 |
NM_028017.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800004792.mm.1 |
TC1800000648.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1800004793.mm.1 |
NM_028017.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800004793.mm.1 |
NM_028017.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800004793.mm.1 |
TC1800000648.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1800004794.mm.1 |
NM_028017.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800004794.mm.1 |
NM_028017.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800004794.mm.1 |
TC1800000648.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1800004789.mm.1 |
NM_028017.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800004789.mm.1 |
NM_028017.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800004789.mm.1 |
TC1800000648.mm.1 |
MTA TC |