Assay Details
| Gene Symbol: Napg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028017.1 | NP_082293.1 | 6 - 7 | 428 | 81 | |
| GenBank mRNA | AK010275.1 | - | 6 - 7 | 481 | 81 | |
| AK090379.1 | - | 6 - 7 | 390 | 81 | ||
| AK154573.1 | - | 6 - 7 | 448 | 81 | ||
| AK164149.1 | - | 6 - 7 | 423 | 81 | ||
| AK167372.1 | - | 6 - 7 | 435 | 81 | ||
| BC026977.1 | - | 6 - 7 | 428 | 5362684 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Napg
Genomic Map