Assay Details
| Gene Symbol: Mbd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013594.2 | NP_038622.2 | 10 - 11 | 1229 | 122 | |
| GenBank mRNA | AK004624.2 | - | 10 - 11 | 1230 | 122 | |
| AK032392.1 | - | 2 - 3 | 173 | 122 | ||
| AK032535.1 | - | 7 - 8 | 727 | 122 | ||
| AK156401.1 | - | 11 - 12 | 1220 | 122 | ||
| AK166042.1 | - | 6 - 7 | 549 | 122 | ||
| BC069837.1 | - | 10 - 11 | 1245 | 30435760 | 122 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mbd1
Genomic Map