Assay Details
| Gene Symbol: Pdgfrb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001146268.1 | NP_001139740.1 | 13 - 14 | 2342 | 61 | |
| NM_008809.2 | NP_032835.2 | 13 - 14 | 2339 | 61 | ||
| GenBank mRNA | AK134785.1 | - | 14 - 15 | 2364 | 61 | |
| AK143247.1 | - | 13 - 14 | 2322 | 61 | ||
| AK158145.1 | - | 7 - 8 | 996 | 61 | ||
| BC055311.1 | - | 13 - 14 | 2354 | 30060666 | 61 | |
| DQ979390.1 | - | 7 - 8 | 995 | 61 | ||
| X04367.1 | - | 13 - 14 | 2048 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pdgfrb
Genomic Map