Assay Details
| Gene Symbol: Rnmt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026440.4 | NP_080716.1 | 7 - 8 | 1225 | 154 | |
| GenBank mRNA | AK011300.1 | - | 7 - 8 | 1209 | 154 | |
| AK082331.1 | - | 7 - 8 | 1192 | 154 | ||
| AK129130.1 | - | 6 - 7 | 1162 | 154 | ||
| BC021794.1 | - | 7 - 8 | 1171 | 4208274 | 154 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rnmt
Genomic Map