Assay Details
| Gene Symbol: Rnmt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170953.1 | NP_001164424.1 | 6 - 7 | 1057 | 139 | |
| GenBank mRNA | AK015403.1 | - | 6 - 7 | 1014 | 139 | |
More Information
Set Membership: |
|

Rnmt
Genomic Map