Assay Details
| Gene Symbol: Mc2r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271717.1 | NP_001258646.1 | 1 - 2 | 79 | 115 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001271717.1 transcripts.
Set Membership: |
|

Mc2r
Genomic Map