Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1800011284.mm.1 |
NM_001136090.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800011284.mm.1 |
NM_001289515.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800011284.mm.1 |
NM_001289516.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800011284.mm.1 |
NM_011972.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800011284.mm.1 |
NM_011972.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800011284.mm.1 |
TC1800001541.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1800011286.mm.1 |
NM_001136090.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800011286.mm.1 |
NM_001289515.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800011286.mm.1 |
NM_001289516.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800011286.mm.1 |
NM_011972.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800011286.mm.1 |
NM_011972.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800011286.mm.1 |
TC1800001541.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1800011285.mm.1 |
NM_001136090.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800011285.mm.1 |
NM_001289515.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800011285.mm.1 |
NM_001289516.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800011285.mm.1 |
NM_011972.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800011285.mm.1 |
NM_011972.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800011285.mm.1 |
TC1800001541.mm.1 |
MTA TC |