Assay Details
| Gene Symbol: Ndst1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008306.4 | NP_032332.2 | 4 - 5 | 1403 | 67 | |
| GenBank mRNA | AF049894.1 | - | 4 - 5 | 1249 | 67 | |
| AF074926.1 | - | 4 - 5 | 1249 | 67 | ||
| AK035642.1 | - | 5 - 6 | 1812 | 67 | ||
| AK147282.1 | - | 5 - 6 | 1796 | 67 | ||
| BC066098.1 | - | 4 - 5 | 1403 | 30533342 | 67 | |
| BC079561.1 | - | 4 - 5 | 1403 | 30533342 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ndst1
Genomic Map