Assay Details
| Gene Symbol: Ndst1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008306.4 | NP_032332.2 | 6 - 7 | 1719 | 80 | |
| GenBank mRNA | AF049894.1 | - | 6 - 7 | 1565 | 80 | |
| AF074926.1 | - | 6 - 7 | 1565 | 80 | ||
| AK147282.1 | - | 7 - 8 | 2112 | 80 | ||
| BC066098.1 | - | 6 - 7 | 1719 | 30533342 | 80 | |
| BC079561.1 | - | 6 - 7 | 1719 | 30533342 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ndst1
Genomic Map