Assay Details
| Gene Symbol: Cd74 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042605.1 | NP_001036070.1 | 3 - 4 | 411 | 71 | |
| NM_010545.3 | NP_034675.1 | 3 - 4 | 411 | 71 | ||
| GenBank mRNA | AK002232.1 | - | 3 - 4 | 411 | 71 | |
| AK020607.1 | - | 3 - 4 | 412 | 71 | ||
| AK154096.1 | - | 3 - 4 | 442 | 71 | ||
| BC003476.1 | - | 3 - 4 | 411 | 2650401 | 71 | |
| BC096435.1 | - | 3 - 4 | 422 | 6530802 | 71 | |
| BG148172.1 | - | 1 - 2 | 79 | 71 | ||
| X00496.1 | - | 2 - 3 | 188 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cd74
Genomic Map