Assay Details
| Gene Symbol: Csnk1g3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152809.2 | NP_690022.2 | 11 - 12 | 1932 | 126 | |
| GenBank mRNA | AK032675.1 | - | 5 - 6 | 487 | 126 | |
| AK034842.1 | - | 1 - 2 | 113 | 126 | ||
| BC033601.1 | - | 8 - 9 | 947 | 126 | ||
More Information
Set Membership: |
|

Csnk1g3
Genomic Map