Assay Details
| Gene Symbol: Ssbp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023672.2 | NP_076161.2 | 2 - 3 | 522 | 74 | |
| NM_198438.1 | NP_940840.1 | 2 - 3 | 522 | 74 | ||
| GenBank mRNA | AF170906.1 | - | 2 - 3 | 252 | 74 | |
| AF500117.1 | - | 2 - 3 | 252 | 74 | ||
| AK011509.1 | - | 2 - 3 | 141 | 74 | ||
| AK011853.1 | - | 2 - 3 | 492 | 74 | ||
| AK017711.1 | - | 2 - 3 | 523 | 74 | ||
| AY167987.1 | - | 2 - 3 | 135 | 74 | ||
| BC003430.1 | - | 2 - 3 | 276 | 2646149 | 74 | |
| BC035475.1 | - | 2 - 3 | 465 | 74 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK011509.1 transcripts.
Set Membership: |
|

Ssbp3
Genomic Map