Assay Details
| Gene Symbol: Ssbp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198438.1 | NP_940840.1 | 5 - 6 | 753 | 95 | |
| GenBank mRNA | AF170906.1 | - | 5 - 6 | 483 | 95 | |
| AF500117.1 | - | 5 - 6 | 483 | 95 | ||
| AK041388.1 | - | 2 - 3 | 349 | 95 | ||
| AK153733.1 | - | 2 - 3 | 194 | 95 | ||
| AK211107.1 | - | 3 - 4 | 203 | 95 | ||
| AY167987.1 | - | 5 - 6 | 366 | 95 | ||
| BC003430.1 | - | 5 - 6 | 507 | 2646149 | 95 | |
| BC035475.1 | - | 5 - 6 | 696 | 95 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK041388.1 transcripts.
Set Membership: |
|

Ssbp3
Genomic Map