Assay Details
| Gene Symbol: Echdc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001254754.1 | NP_001241683.1 | 5 - 6 | 555 | 98 | |
| NM_026728.4 | NP_081004.2 | 6 - 7 | 623 | 98 | ||
| GenBank mRNA | AK005030.1 | - | 6 - 7 | 564 | 98 | |
| AK011365.1 | - | 5 - 6 | 501 | 98 | ||
| AK153878.1 | - | 6 - 7 | 598 | 98 | ||
| AK160482.1 | - | 4 - 5 | 326 | 98 | ||
| AK166388.1 | - | 6 - 7 | 595 | 98 | ||
| AK166965.1 | - | 6 - 7 | 565 | 98 | ||
| BC025104.1 | - | 6 - 7 | 569 | 3591648 | 98 | |
| BC025454.1 | - | 2 - 3 | 107 | 5352625 | 98 | |
More Information
Set Membership: |
|

Echdc2
Genomic Map