Assay Details
| Gene Symbol: Rfx5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017395.2 | NP_059091.2 | 2 - 3 | 164 | 61 | |
| GenBank mRNA | AB221679.1 | - | 2 - 3 | 145 | 61 | |
| AF209854.1 | - | 2 - 3 | 145 | 61 | ||
| AK088686.1 | - | 3 - 4 | 216 | 61 | ||
| BB666132.1 | - | 3 - 4 | 190 | 61 | ||
| BB854843.1 | - | 3 - 4 | 257 | 61 | ||
| BC051965.1 | - | 3 - 4 | 201 | 5702640 | 61 | |
| BC057336.1 | - | 3 - 4 | 190 | 61 | ||
| BC075717.1 | - | 4 - 5 | 273 | 61 | ||
| BC118948.2 | - | 2 - 3 | 145 | 40092396 | 61 | |
| BC118949.2 | - | 2 - 3 | 145 | 40092397 | 61 | |
| BY020562.1 | - | 3 - 4 | 226 | 61 | ||
| BY230827.1 | - | 3 - 4 | 186 | 61 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC051965.1 transcripts.
Set Membership: |
|

Rfx5
Genomic Map