Assay Details
| Gene Symbol: Rfx5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017395.2 | NP_059091.2 | 3 - 4 | 250 | 114 | |
| GenBank mRNA | AB221679.1 | - | 3 - 4 | 231 | 114 | |
| AF209854.1 | - | 3 - 4 | 231 | 114 | ||
| AK088686.1 | - | 4 - 5 | 302 | 114 | ||
| BC051965.1 | - | 4 - 5 | 287 | 5702640 | 114 | |
| BC057336.1 | - | 4 - 5 | 276 | 114 | ||
| BC075717.1 | - | 5 - 6 | 359 | 114 | ||
| BC118948.2 | - | 3 - 4 | 231 | 40092396 | 114 | |
| BC118949.2 | - | 3 - 4 | 231 | 40092397 | 114 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC051965.1 transcripts.
Set Membership: |
|

Rfx5
Genomic Map