Assay Details
| Gene Symbol: Faf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007983.2 | NP_032009.2 | 17 - 18 | 2046 | 65 | |
| GenBank mRNA | AK053752.1 | - | 17 - 18 | 1981 | 65 | |
| BC065098.1 | - | 17 - 18 | 2046 | 30362116 | 65 | |
| U39643.1 | - | 17 - 18 | 2056 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Faf1
Genomic Map