Assay Details
| Gene Symbol: Faf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007983.2 | NP_032009.2 | 1 - 2 | 448 | 92 | |
| GenBank mRNA | AK053752.1 | - | 1 - 2 | 382 | 92 | |
| BC065098.1 | - | 1 - 2 | 448 | 30362116 | 92 | |
More Information
Set Membership: |
|

Faf1
Genomic Map