Assay Details
| Gene Symbol: Pgd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081274.1 | NP_001074743.1 | 6 - 7 | 631 | 82 | |
| GenBank mRNA | AK002894.1 | - | 6 - 7 | 593 | 82 | |
| AK077110.1 | - | 6 - 7 | 593 | 82 | ||
| AK145602.1 | - | 6 - 7 | 595 | 82 | ||
| AK150210.1 | - | 6 - 7 | 594 | 82 | ||
| AK150490.1 | - | 6 - 7 | 590 | 82 | ||
| AK151380.1 | - | 6 - 7 | 590 | 82 | ||
| AK151993.1 | - | 6 - 7 | 590 | 82 | ||
| AK152758.1 | - | 6 - 7 | 590 | 82 | ||
| AK152894.1 | - | 6 - 7 | 590 | 82 | ||
| AK153409.1 | - | 6 - 7 | 587 | 82 | ||
| AK155027.1 | - | 6 - 7 | 590 | 82 | ||
| AK166733.1 | - | 6 - 7 | 585 | 82 | ||
| AK166947.1 | - | 6 - 7 | 567 | 82 | ||
| AK167215.1 | - | 6 - 7 | 585 | 82 | ||
| AK168251.1 | - | 6 - 7 | 590 | 82 | ||
| BC008646.1 | - | 6 - 7 | 562 | 3155147 | 82 | |
| BC011329.1 | - | 6 - 7 | 580 | 3984654 | 82 | |
| BC014793.1 | - | 6 - 7 | 599 | 3155100 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pgd
Genomic Map