Assay Details
| Gene Symbol: Dffb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007859.4 | NP_031885.3 | 4 - 5 | 611 | 100 | |
| GenBank mRNA | AB009377.1 | - | 4 - 5 | 519 | 100 | |
| AK077619.1 | - | 4 - 5 | 657 | 100 | ||
| AK089267.1 | - | 4 - 5 | 588 | 100 | ||
| BC053052.1 | - | 4 - 5 | 541 | 6403143 | 100 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dffb
Genomic Map