Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1800007560.mm.1 |
NM_001142731.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800007560.mm.1 |
NM_001142731.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800007560.mm.1 |
NM_134112.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800007560.mm.1 |
NM_134112.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800007560.mm.1 |
TC1800001012.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1800007561.mm.1 |
NM_001142731.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800007561.mm.1 |
NM_001142731.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800007561.mm.1 |
TC1800001012.mm.1 |
MTA TC |