Assay Details
| Gene Symbol: Ttll10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029264.2 | NP_083540.1 | 12 - 13 | 1481 | 62 | |
| GenBank mRNA | AK133079.1 | - | 12 - 13 | 1482 | 62 | |
| AM690754.1 | - | 9 - 10 | 1365 | 62 | ||
| BC137726.1 | - | 2 - 3 | 305 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ttll10
Genomic Map