Assay Details
| Gene Symbol: Noc2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021303.2 | NP_067278.2 | 10 - 11 | 1243 | 69 | |
| GenBank mRNA | AK076287.1 | - | 9 - 10 | 1285 | 69 | |
| AK078560.1 | - | 10 - 11 | 1219 | 69 | ||
| AK133834.1 | - | 10 - 11 | 1243 | 69 | ||
| AK157765.1 | - | 10 - 11 | 1236 | 69 | ||
| AK164071.1 | - | 3 - 4 | 3459 | 69 | ||
| BC020013.1 | - | 10 - 11 | 1214 | 3663482 | 69 | |
| BC034650.1 | - | 1 - 2 | 807 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Noc2l
Genomic Map