Assay Details
Gene Symbol: Noc2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021303.2 | NP_067278.2 | 11 - 12 | 1382 | 71 | |
GenBank mRNA | AK076287.1 | - | 10 - 11 | 1424 | 71 | |
AK078560.1 | - | 11 - 12 | 1358 | 71 | ||
AK133834.1 | - | 11 - 12 | 1382 | 71 | ||
AK157765.1 | - | 11 - 12 | 1375 | 71 | ||
AK199413.1 | - | 1 - 2 | 172 | 71 | ||
AK210522.1 | - | 1 - 2 | 71 | |||
BC034650.1 | - | 2 - 3 | 946 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
