Assay Details
| Gene Symbol: Mc2r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008560.3 | NP_032586.1 | 1 - 2 | 112 | 71 | |
| GenBank mRNA | AK033100.1 | - | 1 - 2 | 51 | 71 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK033100.1, NM_008560.3 transcripts.
Set Membership: |
|

Mc2r
Genomic Map