Assay Details
| Gene Symbol: Stard6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289648.1 | NP_001276577.1 | 1 - 2 | 48 | 68 | |
| GenBank mRNA | BC087893.1 | - | 1 - 2 | 39 | 6774832 | 68 |
| BY085273.1 | - | 1 - 2 | 49 | 68 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the BC087893.1,NM_001289648.1 transcripts.
This assay is in the 5'UTR region of the BC087893.1,NM_001289648.1 transcripts.
Set Membership: |
|

Stard6
Genomic Map