Assay Details
| Gene Symbol: Slc1a7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146255.2 | NP_666367.2 | 4 - 5 | 720 | 66 | |
| GenBank mRNA | AK044586.1 | - | 4 - 5 | 612 | 66 | |
| AK044671.1 | - | 4 - 5 | 650 | 66 | ||
| BC030400.1 | - | 4 - 5 | 731 | 5363389 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc1a7
Genomic Map