Assay Details
| Gene Symbol: Podn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285958.1 | NP_001272887.1 | 2 - 3 | 330 | 79 | |
| GenBank mRNA | BC052487.1 | - | 2 - 3 | 180 | 79 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001285958.1 transcripts.
Set Membership: |
|

Podn
Genomic Map