Assay Details
| Gene Symbol: Atp6v1g1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK169079.1 | - | 1 - 2 | 454 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Atp6v1g1| Gene Symbol: Atp6v1g1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK169079.1 | - | 1 - 2 | 454 | 62 | |
Set Membership: |
|