Assay Details
| Gene Symbol: Spag16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271533.1 | NP_001258462.1 | 6 - 7 | 667 | 84 | |
| NM_029160.3 | NP_083436.2 | 6 - 7 | 667 | 84 | ||
| GenBank mRNA | AF490391.1 | - | 6 - 7 | 649 | 84 | |
| AK015884.1 | - | 6 - 7 | 668 | 84 | ||
| AK083083.1 | - | 6 - 7 | 646 | 84 | ||
| AK162117.1 | - | 5 - 6 | 582 | 84 | ||
| BC120668.1 | - | 6 - 7 | 666 | 40129591 | 84 | |
| BC145006.1 | - | 6 - 7 | 666 | 9053554 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Spag16
Genomic Map