Assay Details
| Gene Symbol: Spag16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029160.3 | NP_083436.2 | 10 - 11 | 1003 | 92 | |
| GenBank mRNA | AF490391.1 | - | 10 - 11 | 985 | 92 | |
| AK015884.1 | - | 10 - 11 | 1004 | 92 | ||
| BC120668.1 | - | 10 - 11 | 1002 | 40129591 | 92 | |
| BC145006.1 | - | 10 - 11 | 1002 | 9053554 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

Spag16
Genomic Map