Assay Details
| Gene Symbol: Spag16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271533.1 | NP_001258462.1 | 2 - 3 | 197 | 85 | |
| NM_029160.3 | NP_083436.2 | 2 - 3 | 197 | 85 | ||
| GenBank mRNA | AF490391.1 | - | 2 - 3 | 179 | 85 | |
| AK015884.1 | - | 2 - 3 | 198 | 85 | ||
| AK083083.1 | - | 2 - 3 | 176 | 85 | ||
| BC120668.1 | - | 2 - 3 | 196 | 40129591 | 85 | |
| BC145006.1 | - | 2 - 3 | 196 | 9053554 | 85 | |
More Information
Set Membership: |
|

Spag16
Genomic Map