Assay Details
Gene Symbol: Ppp5c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011155.2 | NP_035285.2 | 4 - 5 | 727 | 79 | |
GenBank mRNA | AF018262.1 | - | 4 - 5 | 698 | 79 | |
BC003744.1 | - | 4 - 5 | 683 | 3590322 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
